Learn More
Mopec™ Formaway Waste Collection Container
SDP
Catalog No. 22445410 Shop All Mopec Products
Change view
Click to view available options
Capacity (English):
1 gal.
2.5 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
22-445-410 1 gal.
22-445-351 2.5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-445-410 Supplier Mopec™ Supplier No. BG551
  • $45.25
    $44.65 / Each of 1
    Save $0.60
    1% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Formaway Collection Containers are used to collect spent formalin for neutralizing.
  • Collect waste in a designated collection container
  • Use a funnel with a 'screen insert' while collecting waste to remove tissue debris from waste
  • Add the entire contents of one Formaway bottle to each gallon of aldehyde waste
  • Secure lid on the collection container and agitate container to thoroughly mix solutions, make sure the agitation ultimately results in complete dissolution of Formaway Formalin Neutralizer Powder
  • Allow the mixed solution to stand for 20 to 25 minutes for the reaction to be complete
  • If required, test treated solution for any aldehyde residual prior to disposal (follow instructions for Formaway Formaldehyde Residual Screening Test Kit)
  • Pour treated waste into sanitary sewer and rinse container with cold tap water, preparing it for reuse
Specifications
Capacity (English) 1 gal.
Color White
Autoclavable Not Autoclavable
Material Plastic
Product Type Waste Container