Learn More
Hach Company Stablcal Turbidity Standard, 7500 NTU, Sealed Vial Standard
Catalog No. 501996264 Shop All Hach Company Products
Encompass
Formazin Calibration Standard for use with 2100AN Laboratory Turbidimeter. 7500 NTU ampule.
Catalog No. 50-199-6264 Supplier Hach Company Supplier No. 2584202
Formazin Calibration Standard for use with 2100AN Laboratory Turbidimeter. 7500 NTU ampule.