Learn More
Hach Company Formazin Turbidity Standard, 4000 NTU, 100 mL
Supplier:  Hach Company 246142
Encompass
Turbidity standard can be diluted to be used with any of Hach's turbidimeters TL23, TU52, 2100Q, 2100N, 2100AN series
Catalog No. 50-199-4779