Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic Acid, 10% w/w Volumetric, Reagents

Catalog No. 01335306
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-306 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-306 Supplier Reagents Holdings Llc Supplier No. CF0010001A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Formic Acid, 10% w/w Volumetric, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

pH 1.7
CAS 7732-18-5, 64-18-6
Boiling Point 100°C
Color Colorless
Quantity 1 L
Packaging Poly Bottle
Physical Form Liquid
Molecular Formula H2O
Specific Gravity 1.0242
Synonym Methanoic Acid, Formylic Acid, Aminic Acid
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name water
Molecular Weight (g/mol) 18.02
Percent Purity 10%
Grade Reagent
Chemical Name or Material Formic Acid, 10% w/w Volumetric
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.