Learn More
Sigma Aldrich Fine Chemicals Biosciences Formic acid, 64-18-6, MFCD00003297, 500 mL
Catalog No. NC0850080
Encompass
Linear Formula: HCOOH, Molecular Weight: 46.03, puriss., meets analytical specifications of DAC, FCC, 98.0-100%.
Catalog No. NC0850080 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 27001500MLR
Linear Formula: HCOOH, Molecular Weight: 46.03, puriss., meets analytical specifications of DAC, FCC, 98.0-100%.