Learn More
Sigma Aldrich Fine Chemicals Biosciences FORMIC ACID 98-100%,1L
Catalog No. NC2208840
Encompass
Formic acid 98-100% | CAS No: 64-18-6 | Linear Formula: HCOOH | Molecular Weight: 46.03 | 1 liter
This item is not returnable. View return policy
Catalog No. NC2208840 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1116701000
Formic acid 98-100% | CAS No: 64-18-6 | Linear Formula: HCOOH | Molecular Weight: 46.03 | 1 liter