Learn More
LAB ALLEY Formic Acid 99% LC/MS Grade
SDP
Catalog No. NC3627196
Encompass
Formic Acid is the simplest carboxylic acid containing only one carbon. It is a colorless liquid with a pungent aroma. It is about 10 times stronger than Acetic Acid. The high acidity level and reducing nature of this compound makes it different from other carboxylic acids. LC/MS is a versatile technique that combines the physical separation property of Liquid Chromatography with the mass analysis property of Mass Spectroscopy. LC-MS Grade Formic Acid is high-purity solvent for preparing elution solvents for HPLC separations of protein and peptides. LC/MS Grade Formic Acid is well suited as a chromatography additive for use in LCMS and HPLC applications.
Catalog No. NC3627196 Supplier LAB ALLEY Supplier No. C3994100ML
Formic Acid is the simplest carboxylic acid containing only one carbon. It is a colorless liquid with a pungent aroma. It is about 10 times stronger than Acetic Acid. The high acidity level and reducing nature of this compound makes it different from other carboxylic acids. LC/MS is a versatile technique that combines the physical separation property of Liquid Chromatography with the mass analysis property of Mass Spectroscopy. LC-MS Grade Formic Acid is high-purity solvent for preparing elution solvents for HPLC separations of protein and peptides. LC/MS Grade Formic Acid is well suited as a chromatography additive for use in LCMS and HPLC applications.