Learn More
Proteochem Inc Formic Acid Ampules 10X1Ml (10/Pk)
Catalog No. NC9024207
Encompass_Preferred
Proteomics Grade Formic Acid Ampules 10 x 1.0 mL (CAS 64-18-6) recommended as an HPLC and LCMS mobile phase additive. Each Formic Acid ampule contains 1.0 mL of Proteomics Grade Formic Acid.
Catalog No. NC9024207 Supplier Proteochem Inc Supplier No. LC620110AMPS
Proteomics Grade Formic Acid Ampules 10 x 1.0 mL (CAS 64-18-6) recommended as an HPLC and LCMS mobile phase additive. Each Formic Acid ampule contains 1.0 mL of Proteomics Grade Formic Acid.