Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic Acid C-d 98atom%D 95.0+%, TCI America™
SDP

Catalog No. F11595G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F11595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F11595G Supplier TCI America Supplier No. F11595G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Specifications

Physical Form Clear Liquid at 20°C
CAS 917-71-5
Molecular Formula CH2O2
Quantity 5 g
MDL Number MFCD00059016
Color Colorless
UN Number 1779
InChI Key BDAGIHXWWSANSR-MICDWDOJSA-N
SMILES C(=O)O
IUPAC Name deuterioformic acid
Molecular Weight (g/mol) 47.031
PubChem CID 11263466
Formula Weight 47.03
Percent Purity ≥95.0% (GC)
Chemical Name or Material Formic Acid C-d 98atom%D
Show More Show Less
EINECSNumber (2)-0670
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.