Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic Acid Hydrazide 98%, Thermo Scientific™

Catalog No. AC173251000
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC173251000 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC173251000 Supplier Thermo Scientific Chemicals Supplier No. 173251000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 624-84-0
Infrared Spectrum Authentic
Molecular Formula CH4N2O
Linear Formula HCONHNH2
Quantity 100g
Beilstein 02, 93
Synonym formylhydrazine, formic acid hydrazide, formic hydrazide, hydrazinecarboxaldehyde, formhydrazide, n-formylhydrazine, carbazaldehyde, formylhydrazide, hydrazine, formyl, formal hydrazine
InChI Key XZBIXDPGRMLSTC-UHFFFAOYSA-N
SMILES C(=O)NN
IUPAC Name formohydrazide
Molecular Weight (g/mol) 60.056
PubChem CID 12229
Formula Weight 60.06
Percent Purity ≥97.5%
Chemical Name or Material Formic acid hydrazide, 98%
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 210-867-9
RTECSNumber LQ8615000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.