Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic Acid O-d 98.0atom%D, TCI America™
SDP

Catalog No. F02471G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F02471G Supplier TCI America Supplier No. F02471G

Chemical Identifiers

CAS 925-94-0
Molecular Formula CH2O2
Molecular Weight (g/mol) 47.031
MDL Number MFCD00059017
InChI Key BDAGIHXWWSANSR-DYCDLGHISA-N
PubChem CID 12220501
IUPAC Name deuterio formate
SMILES C(=O)O

Specifications

Physical Form Clear Liquid at 20°C
Quantity 1 g
Color Yellow
UN Number 1779
Formula Weight 47.03
Chemical Name or Material Formic Acid O-d 98.0atom%D
EINECSNumber (2)-0670
RTECSNumber LQ4900000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.