Learn More
IMEB, Inc Tek-Select Formic Acid Plus, 32 oz Bottle, This Formula is a Fixative and Decalcifier, Fresh Tissue can be Placed Directly into Formic Acid Plus to Fix and Decalcify Simultaneously
SDP
Catalog No. 501689479
Encompass_Preferred
Tek-Select Formic Acid Plus, 32 oz Bottle. This formula is a fixative and decalcifier. Fresh tissue can be placed directly into Formic Acid Plus to fix and decalcify simultaneously.
Catalog No. 50-168-9479 Supplier IMEB, Inc Supplier No. FAP1
Tek-Select Formic Acid Plus, 32 oz Bottle. This formula is a fixative and decalcifier. Fresh tissue can be placed directly into Formic Acid Plus to fix and decalcify simultaneously.