Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Formic Acid Triethylamine Complex 5:2

Catalog No. 0656150ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-561-50ML 50 mL
06-561-250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 06-561-50ML Supplier Sigma Aldrich Supplier No. 0656150ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Find details here.Reductant for iridium-catalyzed greener amine synthesis by transfer hydrogenation of imines.A Versatile Catalyst for Reductive Amination by Transfer HydrogenationActivated form of formic acid used for reduction reactions

Specifications

CAS 15077-13-1
Molecular Formula CH2O2 -+ 0.4C6H15N
Refractive Index n20/D 1.428
Linear Formula 5HCOOH -+ 2N(CH2CH3)3
MDL Number MFCD00040600
Quantity 50 mL
Synonym Triethylammonium formate 2:5
Molecular Weight (g/mol) 86.5 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.