Learn More
StatLab™ Formical-4 Decalcifier
GSA_VASDP
Catalog No. STL121432
Change view
Click to view available options
Quantity:
4 x 1 gal.
4 x 32 oz. Bottle
5 gal. Cube
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STL121432 4 x 32 oz. Bottle
STL12141 4 x 1 gal.
STL12145 5 gal. Cube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STL121432 Supplier StatLab™ Supplier No. 121432
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Decalcifies large specimens, allowing better penetration of formaldehyde.
  • Fresh tissue can be placed directly into Formical-4 to fix and decalcify simultaneously
  • Formulated for use on large dense samples that normally take several days to decalcify
  • Works well for IHC and ISH
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 4 x 32 oz. Bottle