Learn More
Medchemexpress LLC Formimidamide (acetate) | 3473-63-0 | ≥97.0% | C3H8N2O2 | 500 G
SDP
Catalog No. 5000434391
Encompass_Preferred
Formimidamide acetate is a biochemical reagent suitable for life science research. It serves as an intermediate in the synthesis of active pharmaceutical ingredients and as a condensing agent for the preparation of pyrimidine and imidazole heterocycles.
  • Used as a biochemical reagent
  • Applicable for life science related research
  • Intermediate in active pharmaceutical ingredient synthesis
  • Condensing agent for pyrimidine and imidazole heterocycles
Catalog No. 50-004-34391 Supplier Medchemexpress LLC Supplier No. HY75153500G
Formimidamide acetate is a biochemical reagent suitable for life science research. It serves as an intermediate in the synthesis of active pharmaceutical ingredients and as a condensing agent for the preparation of pyrimidine and imidazole heterocycles.
  • Used as a biochemical reagent
  • Applicable for life science related research
  • Intermediate in active pharmaceutical ingredient synthesis
  • Condensing agent for pyrimidine and imidazole heterocycles