Learn More
AdipoGen Formononetin
SDP
Catalog No. 502560853
Encompass_Preferred
Chemical. CAS 485-72-3. Formula C16H12O4. MW 268.26. Formononetin is an isoflavonoid phytoestrogenic compound found in soy-based foods and is the precursor of daidzein. It acts as an agonist of the aryl hydrocarbon receptor and a selective inhibitor of ADH gamma the gamma-isoform of alcohol dehydrogenase. It displays potent antitumor, antiviral, anti-inflammatory inhibits NLRP3 inflammasome, antioxidant, neuroprotective and neuroinflammatory properties. Also shown to have vasorelaxant, antihypertensive and antihyperglycemic AMPK activator properties. Induces cell cycle arrest and apoptosis in several cancer cells in vitro and in vivo and shows anti-angiogenic FGFR2 inhibitor, anti-invasive and anti-migratory effects. Increases adipocyte thermogenesis by modulating PPARgamma activity.
Catalog No. 50-256-0853 Supplier AdipoGen Supplier No. CDXF0339M050
Chemical. CAS 485-72-3. Formula C16H12O4. MW 268.26. Formononetin is an isoflavonoid phytoestrogenic compound found in soy-based foods and is the precursor of daidzein. It acts as an agonist of the aryl hydrocarbon receptor and a selective inhibitor of ADH gamma the gamma-isoform of alcohol dehydrogenase. It displays potent antitumor, antiviral, anti-inflammatory inhibits NLRP3 inflammasome, antioxidant, neuroprotective and neuroinflammatory properties. Also shown to have vasorelaxant, antihypertensive and antihyperglycemic AMPK activator properties. Induces cell cycle arrest and apoptosis in several cancer cells in vitro and in vivo and shows anti-angiogenic FGFR2 inhibitor, anti-invasive and anti-migratory effects. Increases adipocyte thermogenesis by modulating PPARgamma activity.