Learn More
Apexbio Technology LLC Formononetin 485-72-3 20mg
SDP
Catalog No. 502240069
Encompass_Preferred
Formononetin (CAS 485-72-3) is an isoflavone primarily derived from red clover (Trifolium pratense) This compound exhibits biological activity against Giardia lamblia by promoting the detachment of trophozoites from the intestinal mucosa thereby disrupting the parasite s adherence and colonization Formononetin is utilized in research investigating anti-parasitic mechanisms host-pathogen interactions in intestinal infections and the broader pharmacological effects of dietary isoflavones Its distinct mode of action supports studies on novel approaches for protozoal infection treatment and the modulation of host cell-pathogen attachment
This item is not returnable. View return policy
Catalog No. 50-224-0069 Supplier Apexbio Technology LLC Supplier No. N162520
Formononetin (CAS 485-72-3) is an isoflavone primarily derived from red clover (Trifolium pratense) This compound exhibits biological activity against Giardia lamblia by promoting the detachment of trophozoites from the intestinal mucosa thereby disrupting the parasite s adherence and colonization Formononetin is utilized in research investigating anti-parasitic mechanisms host-pathogen interactions in intestinal infections and the broader pharmacological effects of dietary isoflavones Its distinct mode of action supports studies on novel approaches for protozoal infection treatment and the modulation of host cell-pathogen attachment