Learn More
Medchemexpress LLC Formononetin | 485-72-3 | 98.79% | 268.26 | 5 MG
SDP
Catalog No. 5000427321
Encompass_Preferred
Formononetin (Standard) is the analytical standard of Formononetin, intended for research and analytical applications. It is a potent FGFR2 inhibitor that potently inhibits angiogenesis and tumor growth. The compound is of analytical standard grade, used as a reference standard in qualitative, quantitative, and methodological research experiments.
  • Analytical standard of Formononetin
  • Potent FGFR2 inhibitor with an IC50 of ~4.31 μM
  • Potently inhibits angiogenesis and tumor growth
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
Catalog No. 50-004-27321 Supplier Medchemexpress LLC Supplier No. HYN0183R5MG
Formononetin (Standard) is the analytical standard of Formononetin, intended for research and analytical applications. It is a potent FGFR2 inhibitor that potently inhibits angiogenesis and tumor growth. The compound is of analytical standard grade, used as a reference standard in qualitative, quantitative, and methodological research experiments.
  • Analytical standard of Formononetin
  • Potent FGFR2 inhibitor with an IC50 of ~4.31 μM
  • Potently inhibits angiogenesis and tumor growth
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS