Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Formononetin | 485-72-3 | MFCD00016948 | 98.8% | 268.26 g/mol | C16H12O4 | 10 MG
SDP

Catalog No. 5000295855
Encompass_Preferred

Analytical standard of the isoflavone formononetin intended for research and analytical applications. Supplied as a 10 mg solid reference material for assay calibration, method development, and compound identification. Not for human or clinical use.

  • High purity: 98.8%.
  • Molecular weight: 268.26 g/mol.
  • CAS number: 485-72-3.
  • Supplied as a small, solid reference material suitable for analytical methods.
  • Intended use: research and analytical applications only.

Catalog No. 50-002-95855 Supplier Medchemexpress LLC Supplier No. HYN0183R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Analytical standard of the isoflavone formononetin intended for research and analytical applications. Supplied as a 10 mg solid reference material for assay calibration, method development, and compound identification. Not for human or clinical use.

  • High purity: 98.8%.
  • Molecular weight: 268.26 g/mol.
  • CAS number: 485-72-3.
  • Supplied as a small, solid reference material suitable for analytical methods.
  • Intended use: research and analytical applications only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.