Learn More
TARGETMOL CHEMICALS INC Formononetin 500MG
SDP
Catalog No. 502880638
Encompass_Preferred
Also available in 1 mL, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Formononetin (Flavosil) is an O-methylated isoflavone and a phytoestrogen from the root of Astragalus membranaceus. Purity 99.38%
Catalog No. 50-288-0638 Supplier TARGETMOL CHEMICALS INC Supplier No. T0724500MG
Also available in 1 mL, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Formononetin (Flavosil) is an O-methylated isoflavone and a phytoestrogen from the root of Astragalus membranaceus. Purity 99.38%