Learn More
Selleck Chemical LLC Formononetin 10mg 485-72-3 Formononetol, Biochanin B
SDP
Supplier:  Selleck Chemical LLC S229910MG
Encompass_Preferred
Formononetin (Formononetol, Biochanin B) is a phytoestrogen from the root of Astragalus membranaceus and an O-methylated isoflavone. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-557-6