Learn More
Cayman Chemical Formononetin, Purity: ≥98%, CAS: 485-72-3, 100 mg
SDP
Catalog No. 502966438
Encompass_Preferred
An isoflavonoid phytoestrogenic compound found in soy-based foods; acts as an agonist of the aryl hydrocarbon receptor (EC50 = 0.13 µM) and has been reported to possess antitumor and antiviral activity
Catalog No. 50-296-6438 Supplier Cayman Chemical Supplier No. 17558100 MG
An isoflavonoid phytoestrogenic compound found in soy-based foods; acts as an agonist of the aryl hydrocarbon receptor (EC50 = 0.13 µM) and has been reported to possess antitumor and antiviral activity