Learn More
Medchemexpress LLC HY-N2389 5mg Medchemexpress, Formosanin C CAS:50773-42-7 Purity:>98%
SDP
Catalog No. 501970390
Encompass_Preferred
Medchemexpress, HY-N2389 5mg Formosanin C CAS:50773-42-7 Formosanin C is a diosgenin saponin isolated from Paris formosana Hayata and an immunomodulator with antitumor activity. Formosanin C induces apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0390 Supplier Medchemexpress LLC Supplier No. HYN23895MG
Medchemexpress, HY-N2389 5mg Formosanin C CAS:50773-42-7 Formosanin C is a diosgenin saponin isolated from Paris formosana Hayata and an immunomodulator with antitumor activity. Formosanin C induces apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.