Learn More
Cayman Chemical FormosnIn C 10mg
SDP
Catalog No. 502945421
Encompass_Preferred
A diosgenin saponin with diverse biological activities; active against C. albicans (MIC = 1 UG/ml) and inhibits C. albicans biofilm formation (MIC = 25 UG/ml); inhibits LPS-induced production of PGE2 and NO in RAW 264.7 macrophages from 0.625-5 µM; induces lipid peroxidation and ferroptosis in MDA-MB-231 cells at 5 µM; delays disease onset in a guinea pig model of S-antigen-induced EAU at 1.5 mg/kg
Catalog No. 50-294-5421 Supplier Cayman Chemical Supplier No. 4023610 MG
A diosgenin saponin with diverse biological activities; active against C. albicans (MIC = 1 UG/ml) and inhibits C. albicans biofilm formation (MIC = 25 UG/ml); inhibits LPS-induced production of PGE2 and NO in RAW 264.7 macrophages from 0.625-5 µM; induces lipid peroxidation and ferroptosis in MDA-MB-231 cells at 5 µM; delays disease onset in a guinea pig model of S-antigen-induced EAU at 1.5 mg/kg