Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Formoterol fumarate | 43229-80-7 | 100.0% | 402.44 | C19H24N2O4 · 1/2 C4H4O4 | 10 MG
SDP

Catalog No. 5000299286
Encompass_Preferred

Formoterol fumarate is the fumarate salt of formoterol, a potent, selective, long-acting β2-adrenoceptor agonist supplied for laboratory research. Consult safety data and product documentation for handling, storage, and disposal instructions.

  • Suitable for research use only
  • High purity, approximately 99.8-99.95%
  • Molecular weight 402.44
  • CAS number 43229-80-7
  • Available in small pack sizes for experimental use, including 10 MG

Catalog No. 50-002-99286 Supplier Medchemexpress LLC Supplier No. HYB001010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Formoterol fumarate is the fumarate salt of formoterol, a potent, selective, long-acting β2-adrenoceptor agonist supplied for laboratory research. Consult safety data and product documentation for handling, storage, and disposal instructions.

  • Suitable for research use only
  • High purity, approximately 99.8-99.95%
  • Molecular weight 402.44
  • CAS number 43229-80-7
  • Available in small pack sizes for experimental use, including 10 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.