Learn More
Apexbio Technology LLC Formoterol fumarate 43229-80-7 50mg
SDP
Catalog No. 5000092561
Encompass_Preferred
Formoterol fumarate (CAS 43229-80-7) is a long-acting 2-adrenergic receptor agonist that selectively binds to 2-adrenergic receptors on bronchial smooth muscle inducing relaxation and bronchodilation Through this mechanism it modulates airway smooth muscle tone leading to sustained improvement in airflow This compound is widely utilized in research exploring pathways of bronchoconstriction receptor signaling and the pharmacological management of obstructive airway diseases such as asthma and chronic obstructive pulmonary disease (COPD)
Catalog No. 50-000-92561 Supplier Apexbio Technology LLC Supplier No. B135950
Formoterol fumarate (CAS 43229-80-7) is a long-acting 2-adrenergic receptor agonist that selectively binds to 2-adrenergic receptors on bronchial smooth muscle inducing relaxation and bronchodilation Through this mechanism it modulates airway smooth muscle tone leading to sustained improvement in airflow This compound is widely utilized in research exploring pathways of bronchoconstriction receptor signaling and the pharmacological management of obstructive airway diseases such as asthma and chronic obstructive pulmonary disease (COPD)