Learn More
Medchemexpress LLC Formoterol fumarate | 43229-80-7 | 99.8% | C19H24N2O4·1/2C4H4O4 | 50 MG
SDP
Catalog No. 5000432257
Encompass_Preferred
Formoterol fumarate is a selective β2-adrenoceptor agonist. This compound abolishes contraction induced by Acetylcholine in bronchioles and can be utilized for research into chronic obstructive pulmonary disease and bronchial asthma.
  • Selective β2-adrenoceptor agonist
  • Abolishes contraction induced by Acetylcholine in bronchioles
  • Can be used for research of chronic obstructive pulmonary disease and bronchial asthma
  • Appears as a white to off-white solid
  • Soluble in DMSO at ≥ 50 mg/mL
Catalog No. 50-004-32257 Supplier Medchemexpress LLC Supplier No. HYB001050MG
Formoterol fumarate is a selective β2-adrenoceptor agonist. This compound abolishes contraction induced by Acetylcholine in bronchioles and can be utilized for research into chronic obstructive pulmonary disease and bronchial asthma.
  • Selective β2-adrenoceptor agonist
  • Abolishes contraction induced by Acetylcholine in bronchioles
  • Can be used for research of chronic obstructive pulmonary disease and bronchial asthma
  • Appears as a white to off-white solid
  • Soluble in DMSO at ≥ 50 mg/mL