Learn More
Medchemexpress LLC Formoterol fumarate | 43229-80-7 | >99.9% | C19H24N2O4 . 1/2C4H4O4 | 1 ML
SDP
Catalog No. 5000432904
Encompass_Preferred
Formoterol fumarate is a selective β2-adrenoceptor agonist, demonstrating similar β2-adrenoceptor selectivity to Salbutamol and Terbutaline. This compound is known for its ability to abolish the contraction induced by Acetylcholine in bronchioles. It is commonly utilized in research applications for conditions such as chronic obstructive pulmonary disease and bronchial asthma.
  • Selective β2-adrenoceptor agonist
  • Abolishes acetylcholine-induced contraction in bronchioles
  • Used in research for chronic obstructive pulmonary disease
  • Utilized in research for bronchial asthma
Catalog No. 50-004-32904 Supplier Medchemexpress LLC Supplier No. HYB001010MM/1ML
Formoterol fumarate is a selective β2-adrenoceptor agonist, demonstrating similar β2-adrenoceptor selectivity to Salbutamol and Terbutaline. This compound is known for its ability to abolish the contraction induced by Acetylcholine in bronchioles. It is commonly utilized in research applications for conditions such as chronic obstructive pulmonary disease and bronchial asthma.
  • Selective β2-adrenoceptor agonist
  • Abolishes acetylcholine-induced contraction in bronchioles
  • Used in research for chronic obstructive pulmonary disease
  • Utilized in research for bronchial asthma