Learn More
Medchemexpress LLC Formoterol fumarate | 43229-80-7 | 99.95% | C19H24N2O4 . 1/2C4H4O4 | 25 MG
SDP
Catalog No. 5000359032
Encompass_Preferred
Formoterol fumarate is a selective β2-adrenoceptor agonist that potently and completely relaxes acetylcholine-pre-contracted human isolated segmental bronchi with a pEC50 of 8.76. It is at least as β2-adrenoceptor selective as Salbutamol and Terbutaline, making it suitable for research into chronic obstructive pulmonary disease and bronchial asthma.
  • Selective β2-adrenoceptor agonist
  • Abolishes contraction induced by acetylcholine in bronchioles
  • Supports research into chronic obstructive pulmonary disease and bronchial asthma
  • Demonstrates potent relaxation of acetylcholine-pre-contracted human isolated segmental bronchi
Catalog No. 50-003-59032 Supplier Medchemexpress LLC Supplier No. 25MGHYB0010
Formoterol fumarate is a selective β2-adrenoceptor agonist that potently and completely relaxes acetylcholine-pre-contracted human isolated segmental bronchi with a pEC50 of 8.76. It is at least as β2-adrenoceptor selective as Salbutamol and Terbutaline, making it suitable for research into chronic obstructive pulmonary disease and bronchial asthma.
  • Selective β2-adrenoceptor agonist
  • Abolishes contraction induced by acetylcholine in bronchioles
  • Supports research into chronic obstructive pulmonary disease and bronchial asthma
  • Demonstrates potent relaxation of acetylcholine-pre-contracted human isolated segmental bronchi