Learn More
Selleck Chemical LLC Formoterol Hemifumarate S2020-1g
SDP
Catalog No. 503109400
Encompass_Preferred
Formoterol Hemifumarate (Eformoterol CGP 25827A NSC 299587 YM 08316 Formoterol fumarate) is a potent selective and long-acting 2-adrenoceptor agonist used in the management of asthma and chronic obstructive pulmonary disease(COPD)
Catalog No. 50-310-9400 Supplier Selleck Chemical LLC Supplier No. S20201G
Formoterol Hemifumarate (Eformoterol CGP 25827A NSC 299587 YM 08316 Formoterol fumarate) is a potent selective and long-acting 2-adrenoceptor agonist used in the management of asthma and chronic obstructive pulmonary disease(COPD)