Learn More
Cayman Chemical Formoterol (hemifumarate hydrate), Purity: ≥99%, 50 mg
SDP
Catalog No. 502971191
Encompass_Preferred
A β2-AR agonist; induces cAMP efflux in, and relaxation of, carbachol-precontracted rat tracheal rings at 1 µM; prevents the late asthmatic response, reduces the number of eosinophils and macrophages in BALF, and reduces bronchial reactivity in a guinea pig model of ovalbumin-induced allergic asthma at 10 µg/ml; an inhibitor of ferroptosis that prevents RSL-3- or erastin-induced cell death in in HT-1080 cells in a concentration-dependent manner; scavenges lipid peroxyl radicals in oxidized egg phosphatidylcholine liposomes at 10-100 µM; reduces acetaminophen-induced acute liver injury and hepatic lipid peroxidation in mice at 1 and 5 mg/kg
Catalog No. 50-297-1191 Supplier Cayman Chemical Supplier No. 1558450 MG
A β2-AR agonist; induces cAMP efflux in, and relaxation of, carbachol-precontracted rat tracheal rings at 1 µM; prevents the late asthmatic response, reduces the number of eosinophils and macrophages in BALF, and reduces bronchial reactivity in a guinea pig model of ovalbumin-induced allergic asthma at 10 µg/ml; an inhibitor of ferroptosis that prevents RSL-3- or erastin-induced cell death in in HT-1080 cells in a concentration-dependent manner; scavenges lipid peroxyl radicals in oxidized egg phosphatidylcholine liposomes at 10-100 µM; reduces acetaminophen-induced acute liver injury and hepatic lipid peroxidation in mice at 1 and 5 mg/kg