Learn More
Leica Biosystems Formula "R" (10 x 2lbs)
Supplier:  Leica Biosystems 3801470
Encompass_Preferred
Formula 'R' is a next generation paraffin that enables thin sectioning with minimal compression and all around ease of use.Formula 'R' is a semitransparent compound which can be used both as an infiltration and embedding medium.It has a low melting point (56-57º C, water bath temperature 45-50º C) and is available in 2 packaging options, a 2 lb. poly bag or 4.5 lb. jug. The perforated poly bag is easy to pour and takes up minimal space.The resealable wide mouth plastic jug facilitates pouring as well as disposing of the used paraffin.
Catalog No. 50-255-2442