Learn More
Electron Microscopy Sciences Formvar 15/ 95 Resin, Powder 100 GR
SDP
Supplier:  Electron Microscopy Sciences 15800
Encompass_Preferred
(Glycerol)
Polyvinyl formvar desiccated. Used to prepare Support films. Soluble in ethylene dichloride, chloroform or dioxane.
Catalog No. 50-980-514