Learn More
Ted Pella Inc Formvar/Carbon 300 mesh, Copper approx. grid hole size: 63µm, 50/pk
SDP
Catalog No. NC9967874
Encompass
A Formvar film covered with a "light" layer of carbon. The heat and electrical conducting properties of carbon help to stabilize the Formvar films when exposed to the electron beam. This is a resilient, all-purpose specimen support film, ideal for mounting thin sections and for applications using lower ranges of magnification.
Catalog No. NC9967874 Supplier Ted Pella Inc Supplier No. 01753F
A Formvar film covered with a "light" layer of carbon. The heat and electrical conducting properties of carbon help to stabilize the Formvar films when exposed to the electron beam. This is a resilient, all-purpose specimen support film, ideal for mounting thin sections and for applications using lower ranges of magnification.