Learn More
Electron Microscopy Sciences Gilder Finder Grid F1, 300 Mesh, Copper
SDP
Catalog No. 501927774
Encompass_Preferred
Used to identify specific areas of mesh by an alphabetical or numerical code built into the mesh. 3.05 mm diameter, 0.7 mil (18 um) thickness. The block of 625 (25x25) individual cells is divided into smaller blocks bounded by thicker crenellated bars. Each block of 25 cells has a central alphabet letter from A to Y for identification purposes.
Grid: Copper
Catalog No. 50-192-7774 Supplier Electron Microscopy Sciences Supplier No. G300F1CU
Used to identify specific areas of mesh by an alphabetical or numerical code built into the mesh. 3.05 mm diameter, 0.7 mil (18 um) thickness. The block of 625 (25x25) individual cells is divided into smaller blocks bounded by thicker crenellated bars. Each block of 25 cells has a central alphabet letter from A to Y for identification purposes.
Grid: Copper