Learn More
Electron Microscopy Sciences Formvar Support Film 5-6 nm thick on 2 X 1 mm Slot Grid Copper Grid
SDP
Catalog No. NC0929395
Encompass_Preferred
A thin film of pure formvar resin coats the shiny side of a slotted grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Standard coating is 5 to 6 nm thick.
Grid: Copper
Catalog No. NC0929395 Supplier Electron Microscopy Sciences Supplier No. FF2010CU
A thin film of pure formvar resin coats the shiny side of a slotted grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Standard coating is 5 to 6 nm thick.
Grid: Copper