Learn More
Electron Microscopy Sciences Formvar Support Film 5-6 nm thick on 2 X 1 mm Slot Grid Nickel Grid
SDP
Catalog No. 5026064
Encompass_Preferred
A thin film of pure formvar resin coats the shiny side of a slotted grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Standard coating is 5 to 6 nm thick.
Grid: Nickel
Catalog No. 50-260-64 Supplier Electron Microscopy Sciences Supplier No. FF2010NI50
A thin film of pure formvar resin coats the shiny side of a slotted grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Standard coating is 5 to 6 nm thick.
Grid: Nickel