Learn More
Electron Microscopy Sciences Formvar Support Film 5-6 nm thick on Square 200 mesh Gold Grid
SDP
Catalog No. 5026059
Encompass_Preferred
Support Film on grids has become a main product line for us since the demand for high quality coated grids has increased. To make your microscopy work easier and to save you a great deal of time we offer you a complete line. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. Standard coating is 5 to 6 nm thick.
Grid: Gold
Catalog No. 50-260-59 Supplier Electron Microscopy Sciences Supplier No. FF200AU50
Support Film on grids has become a main product line for us since the demand for high quality coated grids has increased. To make your microscopy work easier and to save you a great deal of time we offer you a complete line. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. Standard coating is 5 to 6 nm thick.
Grid: Gold