Learn More
Electron Microscopy Sciences Formvar Support Film 5-6 nm thick on London Finder LF200 Gold Grid
SDP
Catalog No. 501900649
Encompass_Preferred
A thin film of pure formvar resin coats the shiny side of a London finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Standard coating is 5 to 6 nm thick.
Grid: Gold
Catalog No. 50-190-0649 Supplier Electron Microscopy Sciences Supplier No. FFLF200AU25
A thin film of pure formvar resin coats the shiny side of a London finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Standard coating is 5 to 6 nm thick.
Grid: Gold