Learn More
Electron Microscopy Sciences Formvar Support Film 20-30 nm thick on Hexagonal 100 mesh Gold Grid
SDP
Catalog No. 501902889
Encompass_Preferred
A thin film of pure formvar resin coats the shiny side of a hexagonal grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Extra thick coating is 20 to 30 nm.
Grid: Gold
Catalog No. 50-190-2889 Supplier Electron Microscopy Sciences Supplier No. FF100HAUET
A thin film of pure formvar resin coats the shiny side of a hexagonal grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Extra thick coating is 20 to 30 nm.
Grid: Gold