Learn More
Electron Microscopy Sciences Formvar Support Film 5-6 nm thick on Hexagonal 100 mesh Nickel Grid
SDP
Catalog No. 501900663
Encompass_Preferred
A thin film of pure formvar resin coats the shiny side of a hexagonal grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Standard coating is 5 to 6 nm thick.
Grid: Nickel
Catalog No. 50-190-0663 Supplier Electron Microscopy Sciences Supplier No. FF100HNI25
A thin film of pure formvar resin coats the shiny side of a hexagonal grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Standard coating is 5 to 6 nm thick.
Grid: Nickel