Learn More
Electron Microscopy Sciences Formvar Support Film 3-4 nm thick on Hexagonal 100 mesh Nickel Grid
SDP
Catalog No. 501902884
Encompass_Preferred
A thin film of pure formvar resin coats the shiny side of a hexagonal grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Nickel
Catalog No. 50-190-2884 Supplier Electron Microscopy Sciences Supplier No. FF100HNIUL
A thin film of pure formvar resin coats the shiny side of a hexagonal grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Nickel