Learn More
Electron Microscopy Sciences Formvar Support Film 10 nm thick on Hexagonal 200 mesh Gold Grid
SDP
Catalog No. 501902897
Encompass_Preferred
A thin film of pure formvar resin coats the shiny side of a hexagonal grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Coating is 10 nm thick.
Grid: Gold
Catalog No. 50-190-2897 Supplier Electron Microscopy Sciences Supplier No. FF200HAUTH
A thin film of pure formvar resin coats the shiny side of a hexagonal grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Coating is 10 nm thick.
Grid: Gold