Learn More
Electron Microscopy Sciences Formvar Support Film 5-6 nm thick on Single 200 µm hole Copper Grid
SDP
Catalog No. 501900753
Encompass_Preferred
A thin film of pure formvar resin coats the shiny side of a single hole grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Standard coating is 5 to 6 nm thick.
Grid: Copper
Catalog No. 50-190-0753 Supplier Electron Microscopy Sciences Supplier No. FFGA200CU25
A thin film of pure formvar resin coats the shiny side of a single hole grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Standard coating is 5 to 6 nm thick.
Grid: Copper