Learn More
Electron Microscopy Sciences Formvar Support Film 5-6 nm thick on Single 75 µm hole Copper Grid
SDP
Catalog No. 501900742
Encompass_Preferred
A thin film of pure formvar resin coats the shiny side of a single hole grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Standard coating is 5 to 6 nm thick.
Grid: Copper
Catalog No. 50-190-0742 Supplier Electron Microscopy Sciences Supplier No. FFGA75CU50
A thin film of pure formvar resin coats the shiny side of a single hole grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Standard coating is 5 to 6 nm thick.
Grid: Copper