Learn More
Electron Microscopy Sciences Formvar Support Film 5-6 nm thick on 2X0.5 mm Slot Grid Gold Grid
SDP
Catalog No. 501900740
Encompass_Preferred
A thin film of pure formvar resin coats the shiny side of a slotted grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Standard coating is 5 to 6 nm thick.
Grid: Gold
Catalog No. 50-190-0740 Supplier Electron Microscopy Sciences Supplier No. FF205AU50
A thin film of pure formvar resin coats the shiny side of a slotted grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Standard coating is 5 to 6 nm thick.
Grid: Gold