Learn More
Electron Microscopy Sciences Formvar Support Film 3-4 nm thick on 2X0.5 mm Slot Grid Copper Grid
SDP
Catalog No. 501902989
Encompass_Preferred
A thin film of pure formvar resin coats the shiny side of a slotted grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Copper
Catalog No. 50-190-2989 Supplier Electron Microscopy Sciences Supplier No. FF205CUUL
A thin film of pure formvar resin coats the shiny side of a slotted grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Copper