Learn More
Electron Microscopy Sciences Formvar Support Film 3-4 nm thick on 2X1 mm Slot Grid Gold Grid
SDP
Catalog No. 501903004
Encompass_Preferred
A thin film of pure formvar resin coats the shiny side of a slotted grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Gold
Catalog No. 50-190-3004 Supplier Electron Microscopy Sciences Supplier No. FF2010AUUL
A thin film of pure formvar resin coats the shiny side of a slotted grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Gold