Learn More
Electron Microscopy Sciences Formvar Support Film 3-4 nm thick on Square 100 mesh Gold Grid
SDP
Catalog No. 501902803
Encompass_Preferred
Support Film on grids has become a main product line for us since the demand for high quality coated grids has increased. To make your microscopy work easier and to save you a great deal of time we offer you a complete line. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Gold
Catalog No. 50-190-2803 Supplier Electron Microscopy Sciences Supplier No. FF100AUUL
Support Film on grids has become a main product line for us since the demand for high quality coated grids has increased. To make your microscopy work easier and to save you a great deal of time we offer you a complete line. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. Ultra thin coating is 3 to 4 nm thick.
Grid: Gold