Learn More
Electron Microscopy Sciences Formvar Support Film 5-6 nm thick on Thin Bar Hexagonal 200 mesh Copper Grid
SDP
Catalog No. 501900709
Encompass_Preferred
A thin film of pure formvar resin coats the shiny side of a thin bar hexagonal grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Standard coating is 5 to 6 nm thick.
Grid: Copper
Catalog No. 50-190-0709 Supplier Electron Microscopy Sciences Supplier No. FFTH200CU50
A thin film of pure formvar resin coats the shiny side of a thin bar hexagonal grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Standard coating is 5 to 6 nm thick.
Grid: Copper